Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m,α,α,α-Tetrafluoroanisole ≥98.0% (by GC)

Supplier: TCI America

F0569-5G F0569-25G
TCF0569-5GEA 109 USD InStock
TCF0569-5G TCF0569-25G
m,α,α,α-Tetrafluoroanisole ≥98.0% (by GC)
m,α,α,α-Tetrafluoroanisole
CAS Number: 1077-01-6
MDL Number: MFCD00236323
Molecular Formula: C7H4F4O
Molecular Weight: 180.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Specific Gravity (20/20): 1.33


Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 105 °C
Density: 1.33 (20 °C)
MDL Number: MFCD00236323
CAS Number: 1077-01-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/91/31/28969131.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty