Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Furfuryl isopropyl sulfide ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 2-((Isopropylthio)methyl)furan

F0742-25G
TCF0742-25GEA 304 USD InStock
TCF0742-25G
Furfuryl isopropyl sulfide ≥98.0% (by GC)
Furfuryl isopropyl sulphide
CAS Number: 1883-78-9
MDL Number: MFCD00040265
Molecular Formula: C8H12OS
Molecular Weight: 156.24
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 76
Specific Gravity (20/20): 1.02

Other Product Names / Synonyms: 2-(Isopropylthiomethyl)furan


Formula: C₈H₁₂OS
MW: 156.25 g/mol
Boiling Pt: 80 °C
Density: 1.02 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00040265
CAS Number: 1883-78-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/86/28969286.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty