Synonyms:
N-Methyl-4-fluorobenzylamine
F0806-5G
F0806-1G
TCF0806-1GEA
38.57
USD
InStock
TCF0806-1G
TCF0806-5G
4-Fluoro-N-methylbenzylamine ≥97.0% (by GC, titration analysis)
4-Fluoro-N-methylbenzylamine
CAS Number: 405-66-3
MDL Number: MFCD03839848
Molecular Formula: C8H10FN
Molecular Weight: 139.17
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 71
Specific Gravity (20/20): 1.05