Synonyms:
(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
F0855-50MG
F0855-10MG
TCF0855-10MGEA
193.07
USD
InStock
TCF0855-10MG
TCF0855-50MG
Forskolin ≥95.0% (by HPLC)
Forskolin
CAS Number: 66575-29-9
MDL Number: MFCD00082317
Molecular Formula: C22H34O7
Molecular Weight: 410.51
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Melting point (°C): 225
Specific rotation [a]20/D: -17 deg
Lambda max.: 305 nm
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Regenerative Medicine Research Reagents, Diterpenes
Other Product Names / Synonyms: Coleonol