Synonyms:
N-Fmoc-pentafluoro-L-phenylalanine
F0900-200MG
F0900-1G
TCF0900-200MGEA
49
USD
InStock
TCF0900-200MG
TCF0900-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine ≥97.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]pentafluoro-L-phenylalanine
CAS Number: 205526-32-5
MDL Number: MFCD00270612
Molecular Formula: C24H16F5NO4
Molecular Weight: 477.39
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -19 deg (C=1, MeOH)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-pentafluoro-L-phenylalanine