Synonyms:
2-(3-Fluorophenyl)ethylamine
F0930-25G
F0930-5G
TCF0930-25GEA
297.4
USD
InStock
TCF0930-25G
TCF0930-5G
3-Fluorophenethylamine ≥98.0% (by GC, titration analysis)
3-Fluorophenethylamine
CAS Number: 404-70-6
MDL Number: MFCD00075376
Molecular Formula: C8H10FN
Molecular Weight: 139.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 87
Flash Point (°C): 83
Specific Gravity (20/20): 1.07
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Amino-2-(3-fluorophenyl)ethane, 3-Fluorophenethylamine