Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Geniposide ≥95.0% (by HPLC)

Supplier: TCI America
Synonyms: Methyl (1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4aalpha,5,7aalpha-tetrahydrocyclopenta[c]pyran-4-carboxylate(1S)-1alpha-(beta-D-Glucopyranosyloxy)-7-(hydroxymethyl)-1,4a alpha,5,7a alpha-tetrahydrocyclopenta[c]pyran-4-carboxylic Acid M

G0385-100MG G0385-1G
TCG0385-100MGEA 111 USD InStock
TCG0385-100MG TCG0385-1G
Geniposide ≥95.0% (by HPLC)
Geniposide
CAS Number: 24512-63-8
MDL Number: MFCD00016659
Molecular Formula: C17H24O10
Molecular Weight: 388.37
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Melting point (°C): 162
Specific rotation [a]20/D: 15 deg (C=1, EtOH)

Product Categories: Bioscience, Glycosides, Glucose, Glycosides, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Others (Phytochemicals)


Formula: C₁₇H₂₄O₁₀
MW: 388.37 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00016659
CAS Number: 24512-63-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/87/28969987.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty