Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-4-methyl-2-pentanone ≥98.0% (by GC)

Supplier: TCI America
Synonyms: Diacetone alcohol

H0272-500ML
TCH0272-500MLEA 31.14 USD InStock
TCH0272-500ML
4-Hydroxy-4-methyl-2-pentanone ≥98.0% (by GC)
4-Hydroxy-4-methyl-2-pentanone
CAS Number: 123-42-2
MDL Number: MFCD00004471
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Melting point (°C): -44
Flash Point (°C): 58
Specific Gravity (20/20): 0.94
Lambda max.: 249 nm

Other Product Names / Synonyms: 4-Hydroxy-4-methyl-2-pentanone


Formula: (CH₃)₂C(OH)CH₂COCH₃
MW: 116.16 g/mol
Boiling Pt: 168 °C
Melting Pt: –44 °C
Density: 0.94 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00004471
CAS Number: 123-42-2
UN: 1148
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/15/28970315.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty