Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperidin-3-ol ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 3-Hydroxypiperidine3-Piperidinol

H0412-25G
TCH0412-25GEA 101 USD InStock
TCH0412-25G
Piperidin-3-ol ≥98.0% (by GC)
Piperidin-3-ol
CAS Number: 6859-99-0
MDL Number: MFCD00014591
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 62
Flash Point (°C): 100

Other Product Names / Synonyms: 3-Piperidinol


Formula: C₅H₁₁NO
MW: 101.15 g/mol
Melting Pt: 62 °C
Flash Pt: 100 °C
MDL Number: MFCD00014591
CAS Number: 6859-99-0
UN: 3263
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/04/33/28970433.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty