Synonyms:
Perfluoroacetone trihydrate
H0425-25G
TCH0425-25GEA
47.69
USD
InStock
TCH0425-25G
Hexafluoroacetone trihydrate ≥97.0% (by titrimetric analysis)
Hexafluoroacetone trihydrate
CAS Number: 34202-69-2
MDL Number: MFCD00149061
Molecular Formula: C3F6O
Molecular Weight: 166.02
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.69
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Hexafluoro-2-propanone