Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxythiophenol ≥98.0% (by GC)

Supplier: TCI America
Synonyms: m-Hydroxythiophenol3-Hydroxybenzene-1-thiol3-Hydroxybenzenethiol3-Mercaptophenol3-Hydroxyphenyl mercaptan3-Hydroxythiophenol1-Hydroxy-3-mercaptobenzene

H1196-5G
TCH1196-5GEA 110.06 USD InStock
TCH1196-5G
3-Hydroxythiophenol ≥98.0% (by GC)
3-Hydroxythiophenol
CAS Number: 40248-84-8
MDL Number: MFCD01317829
Molecular Formula: C6H6OS
Molecular Weight: 126.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Melting point (°C): 17
Flash Point (°C): 96
Specific Gravity (20/20): 1.24

Other Product Names / Synonyms: 3-Hydroxythiophenol, 3-Mercaptophenol


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 135 °C
Melting Pt: 17 °C
Density: 1.24 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD01317829
CAS Number: 40248-84-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/11/34/28971134.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty