Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Hexane, anhydrous ≥96.0% (by GC)

Supplier: TCI America
Synonyms: HexaneHexanenHexyl hydride

H1197-500ML
TCH1197-500MLEA 39.97 USD InStock
TCH1197-500ML
n-Hexane, anhydrous ≥96.0% (by GC)
n-Hexane
CAS Number: 110-54-3
MDL Number: MFCD00009520
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Melting point (°C): -95
Flash Point (°C): -22
Specific Gravity (20/20): 0.66


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68 °C
Melting Pt: –95 °C
Density: 0.66 (20 °C)
Flash Pt: –22 °C
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/11/35/28971135.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty