Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexachloroplatinic(IV) acid hexahydrate

Supplier: TCI America
Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrateChloroplatinic acid hexahydrateDihydrogen hexachloroplatinate (IV) hexahydrate

H1287-1G
TCH1287-1GEA 176.55 USD InStock
TCH1287-1G
Hexachloroplatinic(IV) acid hexahydrate
Hexachloroplatinic (IV) acid hexahydrate
CAS Number: 18497-13-7
MDL Number: MFCD00149910
Molecular Formula: H2Cl6Pt
Molecular Weight: 409.80
Form: Crystal
Color: Slightly Pale Yellow Red
Melting point (°C): 60

Other Product Names / Synonyms: Chloroplatinic Acid, Hexachloroplatinic Acid


Formula: H₂PtCl₆·6H₂O
MW: 517.9 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00149910
CAS Number: 18497-13-7
UN: 2507
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/12/38/28971238.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty