Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxythiobenzamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Synonyms: 4-Hydroxybenzothioamide

H1308-25G H1308-5G
TCH1308-5GEA 33.5 USD InStock
TCH1308-5G TCH1308-25G
4-Hydroxythiobenzamide ≥98.0% (by HPLC, total nitrogen)
4-Hydroxythiobenzamide
CAS Number: 25984-63-8
MDL Number: MFCD04973332
Molecular Formula: C7H7NOS
Molecular Weight: 153.20
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 192


Formula: C₇H₇NOS
MW: 153.2 g/mol
Melting Pt: 192 °C
MDL Number: MFCD04973332
CAS Number: 25984-63-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/12/80/28971280.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty