Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Hydroxytetraphenylcylclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II)

Supplier: TCI America
Synonyms: Shvo's Catalyst

H1322-100MG
TCH1322-100MGEA 120.59 USD InStock
TCH1322-100MG
1-Hydroxytetraphenylcylclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II)
1-Hydroxytetraphenylcylclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II)
CAS Number: 104439-77-2
MDL Number: MFCD03840556
Molecular Formula: C62H42O6Ru2
Molecular Weight: 1085.15
Form: Crystal
Color: Very Pale Yellow Red
Melting point (°C): 227

Other Product Names / Synonyms: Shvo's Catalyst


Formula: C₆₂H₄₂O₆Ru₂
MW: 1085.15 g/mol
Melting Pt: 227 °C
MDL Number: MFCD03840556
CAS Number: 104439-77-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/13/10/28971310.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty