Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trihexylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: Tri-n-hexylamineTri(hex-1-yl)amine

I0360-25ML
TCI0360-25MLEA 64.09 USD InStock
TCI0360-25ML
Trihexylamine ≥98.0% (by GC, titration analysis)
Trihexylamine
[Reagent for Ion-Pair Chromatography]
CAS Number: 102-86-3
MDL Number: MFCD00009523
Molecular Formula: C18H39N
Molecular Weight: 269.52
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 265
Flash Point (°C): 132
Specific Gravity (20/20): 0.80


Formula: C₁₈H₃₉N
MW: 269.51 g/mol
Boiling Pt: 265 °C
Density: 0.80 (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00009523
CAS Number: 102-86-3
UN: 3082
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/85/28972185.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty