Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodobenzoyl chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 2-Iodobenzoylchlorideo-Iodobenzoyl chloride

I0402-25G I0402-10G
TCI0402-25GEA 125.83 USD InStock
TCI0402-25G TCI0402-10G
2-Iodobenzoyl chloride ≥97.0% (by titrimetric analysis)
2-Iodobenzoyl chloride
CAS Number: 609-67-6
MDL Number: MFCD00001040
Molecular Formula: C7H4ClIO
Molecular Weight: 266.46
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Boiling point (°C): 106
Flash Point (°C): 110
Freezing point (°C): 28


Formula: C₇H₄ClIO
MW: 266.47 g/mol
Boiling Pt: 106 °C
Flash Pt: 110 °C
MDL Number: MFCD00001040
CAS Number: 609-67-6
UN: 3261
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/22/38/28972238.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty