Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Iodobenzylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: (4-Iodophenyl)methanamine

I0831-1G I0831-5G
TCI0831-5GEA 331.81 USD InStock
TCI0831-5G TCI0831-1G
4-Iodobenzylamine ≥97.0% (by GC, titration analysis)
4-Iodobenzylamine
CAS Number: 39959-59-6
MDL Number: MFCD00047933
Molecular Formula: C7H8IN
Molecular Weight: 233.05
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 48


Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 115 °C
Melting Pt: 48 °C
MDL Number: MFCD00047933
CAS Number: 39959-59-6
UN: 3259
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/27/19/28972719.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty