Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isoproturon ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Synonyms: 3-(4-Isopropylphenyl)-1,1-dimethylurea

I0843-25G I0843-5G
TCI0843-25GEA 342.96 USD InStock
TCI0843-25G TCI0843-5G
Isoproturon ≥98.0% (by HPLC, total nitrogen)
Isoproturon
CAS Number: 34123-59-6
MDL Number: MFCD00078684
Molecular Formula: C12H18N2O
Molecular Weight: 206.29
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 158
Flash Point (°C): 100
Lambda max.: 240 nm (H2O)

Product Categories: Bioscience, Herbicide Ingredients
Other Product Names / Synonyms: 3-(4-Isopropylphenyl)-1,1-dimethylurea


Formula: C₁₂H₁₈N₂O
MW: 206.29 g/mol
Melting Pt: 158 °C
Flash Pt: 100 °C
MDL Number: MFCD00078684
CAS Number: 34123-59-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/27/31/28972731.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty