Synonyms:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one
I0882-1G
TCI0882-1GEA
173
USD
InStock
TCI0882-1G
5-Iodo-2'-deoxycytidine ≥98.0% (by HPLC, titration analysis)
5-Iodo-2'-deoxycytidine
CAS Number: 611-53-0
MDL Number: MFCD00038063
Molecular Formula: C9H12IN3O4
Molecular Weight: 353.12
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 185
Specific rotation [a]20/D: 29 deg (C=1, H2O)
Storage Temperature: -20°C