Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium tetrahydroborate ∼3 mol/L in THF

Supplier: TCI America
Synonyms: Lithium borohydride Lithium tetrahydridoborate

L0186-100ML
TCL0186-100MLEA 378.48 USD InStock
TCL0186-100ML
Lithium tetrahydroborate ∼3 mol/L in THF
Lithium tetrahydroborate
CAS Number: 16949-15-8
Molecular Formula: H4BLi
Molecular Weight: 21.78
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -18

Other Product Names / Synonyms: Lithium Tetrahydridoborate, Lithium Tetrahydroborate


Formula: H₄BLi
Flash Pt: –18 °C
MDL Number: MFCD00011088
CAS Number: 16949-15-8
UN: 1413
ADR: 4.3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/39/28973139.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty