Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium-tert-butanolate ∼10% ∼1 mol/L in THF

Supplier: TCI America
Synonyms: Lithium tert-butoxide

L0253-100ML
TCL0253-100MLEA 112.63 USD InStock
TCL0253-100ML
Lithium-tert-butanolate ∼10% ∼1 mol/L in THF
Lithium tert-butanolate
CAS Number: 1907-33-1
MDL Number: MFCD00050479
Molecular Formula: C4H9LiO
Molecular Weight: 80.06
Form: Clear Liquid
Color: Pale Yellow
Specific Gravity (20/20): 0.89


Formula: C₄H₉LiO
Density: 0.89 (20 °C)
MDL Number: MFCD00050479
CAS Number: 1907-33-1
UN: 3206
ADR: 4.2,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/87/28973187.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty