Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Lactic acid ≥90.0% (by titrimetric analysis)

Supplier: TCI America
Danger
Synonyms: (R)-(-)-2-Hydroxypropanoic acid (R)-(-)-2-Hydroxypropionic acid (R)-(-)-Lactic acid

L0266-25G L0266-5G
TCL0266-5GEA 106.27 USD InStock
TCL0266-5G TCL0266-25G
D-(-)-Lactic acid ≥90.0% (by titrimetric analysis)
D(-)-Lactic acid


Formula: C₃H₆O₃
MW: 90.08 g/mol
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00068313
CAS Number: 10326-41-7
EINECS: 233-713-2
UN: 3265
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/95/28973195.jpg
Order Now
Specification Test Results
Purity/Analysis Method 90.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty