M0729-25G
M0729-10G
TCM0729-25GEA
165.45
USD
InStock
TCM0729-25G
TCM0729-10G
1-Methylcyclopentanol ≥98.0% (by GC)
1-Methylcyclopentanol
CAS Number: 1462-03-9
MDL Number: MFCD00001364
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Flash Point (°C): 40
Freezing point (°C): 35