Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methylcaproic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1142-25ML
TCM1142-25MLEA 163.02 USD InStock
TCM1142-25ML
5-Methylcaproic acid ≥98.0% (by GC, titration analysis)
5-Methylcaproic acid
CAS Number: 628-46-6
MDL Number: MFCD00036494
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: 98.0% (T,GC)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 102
Specific Gravity (20/20): 0.91


Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 216 °C
Density: 0.91 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00036494
CAS Number: 628-46-6
UN: 3265
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/49/28974849.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T,GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty