Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylmorpholine-N-oxide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 4-Methylmorpholine 4-oxide4-Methylmorpholine N-Oxide

M2192-25G M2192-5G
TCM2192-25GEA 218.23 USD InStock
TCM2192-25G TCM2192-5G
N-Methylmorpholine-N-oxide ≥98.0% (by titrimetric analysis)
N-Methylmorpholine N-oxide
CAS Number: 7529-22-8
MDL Number: MFCD00005947
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 177
Storage Temperature: 0-10°C


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00005947
CAS Number: 7529-22-8
UN: 3082
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/74/28988174.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty