Synonyms:
1-Aminononane
N0297-5ML
N0297-25ML
TCN0297-25MLEA
159.77
USD
InStock
TCN0297-25ML
TCN0297-5ML
Nonylamine ≥98.0% (by GC, titration analysis)
Nonylamine
CAS Number: 112-20-9
MDL Number: MFCD00008249
Molecular Formula: C9H21N
Molecular Weight: 143.27
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 63
Specific Gravity (20/20): 0.79
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Aminononane