N0758-500G
TCN0758-500GEA
41.18
USD
InStock
TCN0758-500G
Nitrobenzene ≥99.5% (by GC)
Nitrobenzene
CAS Number: 98-95-3
Molecular Formula: C6H5NO2
Molecular Weight: 123.11
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Melting point (°C): 6
Flash Point (°C): 88
Specific Gravity (20/20): 1.20