Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-α,α,α-trifluorotoluene ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 1-Nitro-4-(trifluoromethyl)benzene 4-Nitrobenzotrifluoride

N0902-5G N0902-25G
TCN0902-25GEA 456.75 USD InStock
TCN0902-25G TCN0902-5G
4-Nitro-α,α,α-trifluorotoluene ≥98.0% (by GC)
4-Nitro-α,α,α-trifluorotoluene
CAS Number: 402-54-0
MDL Number: MFCD00007358
Molecular Formula: C7H4F3NO2
Molecular Weight: 191.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 82
Melting point (°C): 42
Flash Point (°C): 88

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Nitro-4-(trifluoromethyl)benzene, 4-Nitro-alpha,alpha,alpha-trifluorotoluene


Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Boiling Pt: 82 °C
Melting Pt: 42 °C
Flash Pt: 88 °C
MDL Number: MFCD00007358
CAS Number: 402-54-0
UN: 3431
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/00/91/28990091.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty