Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalacetic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Oxalacetic acid2-Succinic acid

O0072-5G
TCO0072-5GEA 58.84 USD InStock
TCO0072-5G
Oxalacetic acid ≥97.0% (by titrimetric analysis)
Oxalacetic acid
CAS Number: 328-42-7
MDL Number: MFCD00002592
Molecular Formula: C4H4O5
Molecular Weight: 132.07
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Formula: C₄H₄O₅
MW: 132.07 g/mol
MDL Number: MFCD00002592
CAS Number: 328-42-7
UN: 3261
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/80/28990380.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty