Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Octylthiophene ≥98.0% (by GC)

Supplier: TCI America
Synonyms: 2-n-Octylthiophene

O0376-5G
TCO0376-5GEA 64 USD InStock
TCO0376-5G
2-Octylthiophene ≥98.0% (by GC)
2-Octylthiophene
CAS Number: 880-36-4
MDL Number: MFCD00041015
Molecular Formula: C12H20S
Molecular Weight: 196.35
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 100
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C


Formula: C₁₂H₂₀S
MW: 196.36 g/mol
Boiling Pt: 125 °C
Density: 0.92 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00041015
CAS Number: 880-36-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/05/28990605.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty