Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Pentadiene ≥95.0% (by GC)

Supplier: TCI America

P0805-5G P0805-1G
TCP0805-1GEA 377.41 USD InStock
TCP0805-1G TCP0805-5G
1,2-Pentadiene ≥95.0% (by GC)
1,2-Pentadiene
CAS Number: 591-95-7
MDL Number: MFCD00039943
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 45
Specific Gravity (20/20): 0.70
Storage Temperature: 0-10°C

Other Product Names / Synonyms: Ethylallene


https://us-legacy.vwr.com/stibo/web/std.lang.all/14/71/28991471.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty