Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America
Synonyms: 1,2-Diaminobenzene dihydrochloride 1,2-Phenylenediamine dihydrochloride 1,2-Phenylenediammonium dichloride Benzene-1,2-diamine dihydrochloride

P1144-5G P1144-1G
TCP1144-1GEA 23.45 USD InStock
TCP1144-1G TCP1144-5G
o-Phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research
o-Phenylenediamine dihydrochloride
[for Biochemical Research]
CAS Number: 615-28-1
MDL Number: MFCD00012966
Molecular Formula: C6H8N2
Molecular Weight: 181.06
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 205

Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 1,2-Diaminobenzene Dihydrochloride, OPD.2HCl


Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00012966
CAS Number: 615-28-1
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/17/73/28991773.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty