Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinium(IV) oxide ≥85.0% (by gravimetric analysis)

Supplier: TCI America
Synonyms: Platinum dioxide Adam's type catalyst

P1720-200MG
TCP1720-200MGEA 78.58 USD InStock
TCP1720-200MG
Platinium(IV) oxide ≥85.0% (by gravimetric analysis)
Platinium (IV) oxide
CAS Number: 1314-15-4
MDL Number: MFCD00011184
Molecular Formula: O2Pt
Molecular Weight: 227.08
Purity/Analysis Method: 85.0% (W)
Form: Crystal
Color: Gray


Formula: O₂Pt
MW: 227.08 g/mol
MDL Number: MFCD00011184
CAS Number: 1314-15-4
UN: 1479
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/30/46/28993046.jpg
Order Now
Specification Test Results
Purity/Analysis Method 85.0% (W)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty