Synonyms:
3-Pyridyl Triflate
, Trifluoromethanesulfonic Acid 3-Pyridyl Ester
, Trifluoromethanesulphonic Acid 3-Pyridyl Ester
P1860-5G
TCP1860-5GEA
146.1
USD
InStock
TCP1860-5G
3-Pyridyl trifluoromethanesulfonate ≥98.0% (by GC)
3-Pyridyl trifluoromethanesulfonate
CAS Number: 107658-27-5
MDL Number: MFCD00799579
Molecular Formula: C6H4F3NO3S
Molecular Weight: 227.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.49
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid 3-Pyridyl Ester, 3-Pyridyl Triflate