Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum(II) chloride ≥95.0% (by gravimetric analysis)

Supplier: TCI America
Synonyms: Platinum dichloride

P1920-1G
TCP1920-1GEA 184.85 USD InStock
TCP1920-1G
Platinum(II) chloride ≥95.0% (by gravimetric analysis)
Platinum (II) chloride
CAS Number: 10025-65-7
MDL Number: MFCD00011181
Molecular Formula: PtCl2
Molecular Weight: 265.98
Purity/Analysis Method: 95.0% (W)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 581


Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 °C
MDL Number: MFCD00011181
CAS Number: 10025-65-7
UN: 3260
ADR: 8,III
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (W)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty