Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium perrhenate

Supplier: TCI America

P1939-1G
TCP1939-1GEA 75 USD InStock
TCP1939-1G
Potassium perrhenate
Potassium perrhenate
CAS Number: 10466-65-6
MDL Number: MFCD00011365
Molecular Formula: HReO4
Molecular Weight: 289.30
Form: Crystal
Color: White
Melting point (°C): 550


Formula: KO₄Re
MW: 289.3 g/mol
Melting Pt: 550 °C
MDL Number: MFCD00011365
CAS Number: 10466-65-6
UN: 1479
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/98/28993198.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty