Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pifithrin-mu ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Synonyms: 2-Phenylethynesulfonamide

P2048-10MG P2048-100MG
TCP2048-10MGEA 29.44 USD InStock
TCP2048-10MG TCP2048-100MG
Pifithrin-mu ≥98.0% (by HPLC, total nitrogen)
Pifithrin-mu
CAS Number: 64984-31-2
MDL Number: MFCD00181531
Molecular Formula: C8H7NO2S
Molecular Weight: 181.21
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 137
Storage Temperature: 0-10°C

Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents
Other Product Names / Synonyms: 2-Phenylethynesulfonamide


Formula: C₈H₇NO₂S
MW: 181.21 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00181531
CAS Number: 64984-31-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/32/76/28993276.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty