Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quetiapine hemifumarate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: ICI-204636Seroquel2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol bis[(2E)-2-butenedioate]

Q0092-5G Q0092-25G
TCQ0092-25GEA 673.96 USD InStock
TCQ0092-25G TCQ0092-5G
Quetiapine hemifumarate ≥98.0% (by HPLC, titration analysis)
Quetiapine hemifumarate
CAS Number: 111974-72-2
MDL Number: MFCD03423782
Molecular Formula: C21H25N3O2S
Molecular Weight: 441.55
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 174


Formula: C₂₁H₂₅N₃O₂S
MW: 441.55 g/mol
Melting Pt: 174 °C
MDL Number: MFCD03423782
CAS Number: 111974-72-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/26/28993726.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty