Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tropaeolin O

Supplier: TCI America
Synonyms: Acid Orange 6Sulfo orange2,4-Dihydroxyazobenzene-4'-sulfonic Acid Sodium SaltResorcinol YellowSodium 2,4-Dihydroxyazobenzene-4'-sulfonate

R0010-1G
TCR0010-1GEA 23.45 USD InStock
TCR0010-1G
Tropaeolin O
Tropaeolin O
Form: Crystal
Color: Reddish Yellow
Flash Point (°C): 93


Formula: C₁₂H₁₀N₂O₅S
MW: 316.26 g/mol
Flash Pt: 93 °C
MDL Number: MFCD00007499
CAS Number: 547-57-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/42/28993742.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty