Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rotenone ≥95.0% (by HPLC)

Supplier: TCI America

R0090-5G R0090-25G
TCR0090-25GEA 478 USD InStock
TCR0090-25G TCR0090-5G
Rotenone ≥95.0% (by HPLC)
Rotenone
CAS Number: 83-79-4
MDL Number: MFCD09025614
Molecular Formula: C23H22O6
Molecular Weight: 394.42
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Color: White
Boiling point (°C): 220
Melting point (°C): 183
Specific rotation [a]20/D: -114 deg (C=1, CHCl3)


Formula: C₂₃H₂₂O₆
Boiling Pt: 220 °C
Melting Pt: 183 °C
MDL Number: MFCD09025614
CAS Number: 83-79-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/92/28993792.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty