Synonyms:
Pentane
S0277-5ML
TCS0277-5MLEA
104
USD
InStock
TCS0277-5ML
n-Pentane ≥99.5% (by GC)
n-Pentane
[Standard Material for GC]
CAS Number: 109-66-0
MDL Number: MFCD00009498
Molecular Formula: C5H12
Molecular Weight: 72.15
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 37
Flash Point (°C): -49
Freezing point (°C): -129
Specific Gravity (20/20): 0.63