S0280-5ML
TCS0280-5MLEA
92.37
USD
InStock
TCS0280-5ML
n-Octane ≥99.5% (by GC)
n-Octane
[Standard Material for GC]
CAS Number: 111-65-9
MDL Number: MFCD00009556
Molecular Formula: C8H18
Molecular Weight: 114.23
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 125
Melting point (°C): -57
Flash Point (°C): 14
Specific Gravity (20/20): 0.70