S0281-5ML
TCS0281-5MLEA
84.25
USD
InStock
TCS0281-5ML
n-Nonane ≥99.5% (by GC)
n-Nonane
[Standard Material for GC]
CAS Number: 111-84-2
MDL Number: MFCD00009574
Molecular Formula: C9H20
Molecular Weight: 128.26
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 150
Melting point (°C): -51
Flash Point (°C): 31
Specific Gravity (20/20): 0.72