S0282-5ML
TCS0282-5MLEA
75.2
USD
InStock
TCS0282-5ML
n-Decane ≥99.5% (by GC)
n-Decane
[Standard Material for GC]
CAS Number: 124-18-5
MDL Number: MFCD00008954
Molecular Formula: C10H22
Molecular Weight: 142.29
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 174
Flash Point (°C): 50
Specific Gravity (20/20): 0.73