Synonyms:
Acetic acid methyl ester
S0300-5ML
TCS0300-5MLEA
129.18
USD
InStock
TCS0300-5ML
Methyl acetate ≥99.5% (by GC)
Methyl acetate
[Standard Material for GC]
CAS Number: 79-20-9
MDL Number: MFCD00008711
Molecular Formula: C3H6O2
Molecular Weight: 74.08
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 57
Flash Point (°C): -16
Specific Gravity (20/20): 0.94
Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard)
Other Product Names / Synonyms: Acetic Acid Methyl Ester