Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentene ≥99.5% (by GC)

Supplier: TCI America
Synonyms: Amylene

S0335-5ML
TCS0335-5MLEA 99.47 USD InStock
TCS0335-5ML
1-Pentene ≥99.5% (by GC)
1-Pentene
[Standard Material for GC]
CAS Number: 109-67-1
MDL Number: MFCD00003567
Molecular Formula: C5H10
Molecular Weight: 70.14
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -51
Storage Temperature: 0-10°C


Formula: C₅H₁₀
MW: 70.13 g/mol
Flash Pt: –51 °C
MDL Number: MFCD00003567
CAS Number: 109-67-1
UN: 1108
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/40/62/28994062.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.5% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty