S0392-25G
TCS0392-25GEA
120.79
USD
InStock
TCS0392-25G
Sodium hexafluorophosphate ≥98.0% (by titrimetric analysis)
Sodium hexafluorophosphate
CAS Number: 21324-39-0
MDL Number: MFCD00011122
Molecular Formula: F6NaP
Molecular Weight: 167.95
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White