Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 1 mol/L in water

Supplier: TCI America
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

S0542-500ML
TCS0542-500MLEA 22.51 USD InStock
TCS0542-500ML
Sodium hydroxide 1 mol/L in water
Sodium hydroxide
CAS Number: 1310-73-2
MDL Number: MFCD00003548
Molecular Formula: HNaO
Molecular Weight: 40.00
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.04


Formula: NaOH
MW: 40 g/mol
Density: 1.04 (20 °C)
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/41/77/28994177.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty