Synonyms:
Iodoacetic Acid N-Succinimidyl EsterN-(Iodoacetoxy)succinimide
S0844-100MG
TCS0844-100MGEA
238.53
USD
InStock
TCS0844-100MG
N-Succinimidyl iodoacetate (SIA) ≥95.0% (by total nitrogen basis)
SIA (N-Succinimidyl iodoacetate)
CAS Number: 39028-27-8
MDL Number: MFCD00058451
Molecular Formula: C6H6INO4
Molecular Weight: 283.02
Purity/Analysis Method: 95.0% (N)
Form: Crystal
Color: White
Melting point (°C): 148
Storage Temperature: 0°C
Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: Iodoacetic Acid N-Succinimidyl Ester, N-(Iodoacetoxy)succinimide